Ethyl difluoroacetate 98%
Leverantör: Apollo Scientific
Fara
Synonyms: Difluoroacetic ethyl ester
PC3200-100G PC3200-25G PC3200-500G
APOSPC3200-100GEA 515 SEK
APOSPC3200-100G APOSPC3200-25G APOSPC3200-500G
Ethyl difluoroacetate 98%
Ethyl difluoroacetate
Formel: C₄H₆F₂O₂
MW: 124,09 g/mol
Kokpunkt: 98…99 °C
Densitet: 1,18 g/cm³ (25 °C)
Flampunkt: 23,9 °C
Storage Temperature: Omgivande
MDL Number: MFCD00013578
CAS nummer: 454-31-9
EINECS: 207-223-4
UN: 2924
ADR: 3+8,III