4-(Trifluoromethyl)cyclohexanol (cis- and trans- mixture)
Leverantör: Apollo Scientific
Varning
Synonyms: 4-(Trifluoromethyl)cyclohexanol, 4-(Trifluoromethyl)cyclohexan-1-ol
PC31341-1G PC31341-5G PC31341-10G
APOSPC31341-1GEA 670 SEK
APOSPC31341-1G APOSPC31341-5G APOSPC31341-10G
4-(Trifluoromethyl)cyclohexanol (cis- and trans- mixture)
4-(Trifluoromethyl)cyclohexanol (cis- and trans- mixture)
Formel: C₇H₁₁F₃O
MW: 168,16 g/mol
Kokpunkt: 70…75 °C/15mmHg
Densitet: 1,23 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00102144
CAS nummer: 30129-18-1