1H,1H,7H-Dodecafluoroheptyl acrylate 97%
Leverantör: Apollo Scientific
Varning
Synonyms: DFHA, Acrylic acid 2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoroheptyl ester, 2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoroheptyl Acrylate
PC3132-100G PC3132-25G PC3132-5G
APOSPC3132-100GEA 0 SEK
APOSPC3132-100G APOSPC3132-25G APOSPC3132-5G
1H,1H,7H-Dodecafluoroheptyl acrylate 97%
1H,1H,7H-Dodecafluoroheptyl acrylate
Formel: C₁₀H₆F₁₂O₂
MW: 386,14 g/mol
Kokpunkt: 197 °C
Densitet: 1,581 g/cm³ (25 °C)
Flampunkt: 101 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00080746
CAS nummer: 2993-85-3
EINECS: 221-064-8