5,7-Bis(trifluoromethyl)-4-hydroxyquinoline 97%
Leverantör: Apollo Scientific
Varning
PC3116G-250MG PC3116G-1G
APOSPC3116G-250MGEA 685 SEK
APOSPC3116G-250MG APOSPC3116G-1G
5,7-Bis(trifluoromethyl)-4-hydroxyquinoline 97%
5,7-Bis(trifluoromethyl)-4-hydroxyquinoline
Formel: C₁₁H₅F₆NO
Storage Temperature: Omgivande
MDL Number: MFCD00153141
CAS nummer: 237076-72-1