3,5-Bis(trifluoromethyl)phenyl Isocyanate 97%
Leverantör: Apollo Scientific
Fara
Synonyms: 1-Isocyanato-3,5-bis(trifluoromethyl)benzene, 3,5-Bis(trifluoromethyl)phenylisocyanate, Isocyanic Acid 3,5-Bis(trifluoromethyl)phenyl Ester
PC3115A-25G PC3115A-5G
APOSPC3115A-25GEA 3325 SEK
APOSPC3115A-25G APOSPC3115A-5G
3,5-Bis(trifluoromethyl)phenyl Isocyanate 97%
3,5-Bis(trifluoromethyl)phenyl Isocyanate
  • May form small amount of sediment.
Formel: C₉H₃F₆NO
MW: 255.12 g/mol
Kokpunkt: 62 °C/2mmHg
Densitet: 1.476 g/cm³ (25 °C)
Flampunkt: 67 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00013559
CAS nummer: 16588-74-2
UN: 2206
ADR: 6.1 III