1-Bromo-3,5-bis(trifluoromethyl)benzene 99%
Leverantör: Apollo Scientific
Varning
Synonyms: 3,5-Bis(trifluoromethyl)-1-bromobenzene, 1-Bromo-3,5-bis(trifluoromethyl)benzene, 5-Bromo-α,α,α,β,β,β-Hexafluoro-m-xylene, 1,3-Bis(trifluoromethyl)-5-bromobenzene, 3,5-Bis(trifluoromethyl)bromobenzene, 5-Bromo-α,α,α,α',α',α'-Hexafluoro-m-xylene
PC3106-100G PC3106-25G PC3106-500G
APOSPC3106-100GEA 465 SEK
APOSPC3106-100G APOSPC3106-25G APOSPC3106-500G
1-Bromo-3,5-bis(trifluoromethyl)benzene 99%
1-Bromo-3,5-bis(trifluoromethyl)benzene
Reagent for adding inductively withdrawing substituent into precursors for carbocations, J.O.C., 1984, 49, 2608
Formel: C₈H₃BrF₆
MW: 293.01 g/mol
Kokpunkt: 155 °C
Smältpunkt: –16 °C
Densitet: 1.699 g/cm³ (25 °C)
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000381
CAS nummer: 328-70-1