2-Bromo-2',5'-difluoroacetophenone, Technical Grade
Leverantör: Apollo Scientific
Fara
PC3057-25G PC3057-5G
APOSPC3057-25GEA 11150 SEK
APOSPC3057-25G APOSPC3057-5G
2-Bromo-2',5'-difluoroacetophenone, Technical Grade
2-Bromo-2',5'-difluoroacetophenone
Formel: C₈H₅BrF₂O
Smältpunkt: 37…40 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00142839
CAS nummer: 258513-41-6
UN: 1759
ADR: 8,III