4-(2,4-Difluorobenzoyl)piperidine hydrochloride
Leverantör: Apollo Scientific
PC305000-100G PC305000-25G
APOSPC305000-100GEA 2175 SEK
APOSPC305000-100G APOSPC305000-25G
4-(2,4-Difluorobenzoyl)piperidine hydrochloride
4-(2,4-Difluorobenzoyl)piperidine hydrochloride
Formel: C₁₂H₁₃F₂NO
MW: 261.70 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD01313310
CAS nummer: 106266-04-0