4-Methoxy-3-methyl-5-(trifluoromethyl)aniline 98%
Leverantör: Apollo Scientific
Varning
Synonyms: Benzenamine, 4-methoxy-3-methyl-5-(trifluoromethyl)-, 4-Methoxy-3-methyl-5-(trifluoromethyl)benzenamine
PC303129-5G PC303129-1G
APOSPC303129-5GEA 4475 SEK
APOSPC303129-5G APOSPC303129-1G
4-Methoxy-3-methyl-5-(trifluoromethyl)aniline 98%
4-Methoxy-3-methyl-5-(trifluoromethyl)aniline
Formel: C₉H₁₀F₃NO
MW: 205.18 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD22682815
CAS nummer: 1431329-84-8
UN: 2810
ADR: 6.1 III