4-n-Butoxy-2,6-difluorobenzoic acid 98%
Leverantör: Apollo Scientific
Synonyms: 4-butoxy-2,6-difluorobenzoic acid
PC303075-10G PC303075-5G PC303075-1G
APOSPC303075-10GEA 3875 SEK
APOSPC303075-10G APOSPC303075-5G APOSPC303075-1G
4-n-Butoxy-2,6-difluorobenzoic acid 98%
4-n-Butoxy-2,6-difluorobenzoic acid
Formel: C₁₁H₁₂F₂O₃
MW: 230,21 g/mol
Smältpunkt: 112…115 °C
Storage Temperature: Omgivande
MDL Number: MFCD22201018
CAS nummer: 123843-56-1