2,4-Difluoro-3-ethoxybenzonitrile 97%
Leverantör: Apollo Scientific
Synonyms: 3-Ethoxy-2,4-difluorobenzonitrile
PC303063-10G PC303063-5G PC303063-1G PC303063-25G PC303063-100G
APOSPC303063-10GEA 7525 SEK
APOSPC303063-10G APOSPC303063-5G APOSPC303063-1G APOSPC303063-25G APOSPC303063-100G
2,4-Difluoro-3-ethoxybenzonitrile 97%
2,4-Difluoro-3-ethoxybenzonitrile
Formel: C₉H₇F₂NO
Storage Temperature: Omgivande
MDL Number: MFCD09258732
CAS nummer: 1017778-32-3