2,6-Dichloro-3-(trifluoromethyl)benzyl bromide 98%
Leverantör: Apollo Scientific
Synonyms: 2-(Bromomethyl)-1,3-dichloro-4-(trifluoromethyl)benzene
PC303019-10G PC303019-5G PC303019-1G PC303019-25G
APOSPC303019-10GEA 5450 SEK
APOSPC303019-10G APOSPC303019-5G APOSPC303019-1G APOSPC303019-25G
2,6-Dichloro-3-(trifluoromethyl)benzyl bromide 98%
2,6-Dichloro-3-(trifluoromethyl)benzyl bromide
Formel: C₈H₄BrCl₂F₃
MW: 307.93 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11519349
CAS nummer: 1092461-07-8