2-Fluoro-5-methyl-4-(trifluoromethyl)benzaldehyde 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 2-fluoro-5-methyl-4-(trifluoromethyl)benzaldehyde
PC302920-5G PC302920-1G
APOSPC302920-5GEA 5050 SEK
APOSPC302920-5G APOSPC302920-1G
2-Fluoro-5-methyl-4-(trifluoromethyl)benzaldehyde 97%
2-Fluoro-5-methyl-4-(trifluoromethyl)benzaldehyde
Formel: C₉H₆F₄O
MW: 206.14 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD19687179
CAS nummer: 1323966-40-0