2-Fluoro-4-(trifluoromethoxy)benzaldehyde
Leverantör: Apollo Scientific
Varning
PC302914-10G PC302914-1G PC302914-5G
APOSPC302914-10GEA 6500 SEK
APOSPC302914-10G APOSPC302914-1G APOSPC302914-5G
2-Fluoro-4-(trifluoromethoxy)benzaldehyde
2-Fluoro-4-(trifluoromethoxy)benzaldehyde
Formel: C₈H₄F₄O₂
MW: 208.11 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD16652440
CAS nummer: 1227628-83-2