4-Methyl-3-(trifluoromethyl)benzaldehyde
Leverantör: Apollo Scientific
Varning
PC302888-5G PC302888-10G PC302888-1G
APOSPC302888-5GEA 6575 SEK
APOSPC302888-5G APOSPC302888-10G APOSPC302888-1G
4-Methyl-3-(trifluoromethyl)benzaldehyde
4-Methyl-3-(trifluoromethyl)benzaldehyde
Formel: C₉H₇F₃O
MW: 188.15 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD04116033
CAS nummer: 93249-45-7