3-Ethoxy-2,4-difluorobenzoic acid
Leverantör: Apollo Scientific
Varning
PC302722-25G PC302722-5G PC302722-1G
APOSPC302722-25GEA 11125 SEK
APOSPC302722-25G APOSPC302722-5G APOSPC302722-1G
3-Ethoxy-2,4-difluorobenzoic acid
3-Ethoxy-2,4-difluorobenzoic acid
Formel: C₉H₈F₂O₃
MW: 202,15 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09258723
CAS nummer: 1017779-88-2