3-Chloro-4-ethoxy-5-fluorobenzylamine 97%
Leverantör: Apollo Scientific
PC302528-1G PC302528-5G
APOSPC302528-1GEA 1875 SEK
APOSPC302528-1G APOSPC302528-5G
3-Chloro-4-ethoxy-5-fluorobenzylamine 97%
3-Chloro-4-ethoxy-5-fluorobenzylamine
Formel: C₉H₁₁ClFNO
MW: 203,64 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09832346
CAS nummer: 1017779-02-0