3-(5-Fluoro-2-(trifluoromethoxy)phenyl)propionic acid 98%
Leverantör: Apollo Scientific
PC302519-1G PC302519-5G
APOSPC302519-1GEA 1750 SEK
APOSPC302519-1G APOSPC302519-5G
3-(5-Fluoro-2-(trifluoromethoxy)phenyl)propionic acid 98%
3-(5-Fluoro-2-(trifluoromethoxy)phenyl)propionic acid
Formel: C₁₀H₈F₄O₃
Storage Temperature: Omgivande
MDL Number: MFCD11519341
CAS nummer: 1092460-96-2