3-(2-Methoxy-5-(trifluoromethoxy)phenyl)propionic acid 98%
Leverantör: Apollo Scientific
PC302502-1G PC302502-5G
APOSPC302502-1GEA 2100 SEK
APOSPC302502-1G APOSPC302502-5G
3-(2-Methoxy-5-(trifluoromethoxy)phenyl)propionic acid 98%
3-(2-Methoxy-5-(trifluoromethoxy)phenyl)propionic acid
Formel: C₁₁H₁₁F₃O₄
Storage Temperature: Omgivande
MDL Number: MFCD09832351
CAS nummer: 1017779-17-7