3-Chloro-4-(trifluoromethyl)benzoic acid 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 3-Chloro-α,α,α-trifluoro-p-toluic acid
PC302395-25G PC302395-5G PC302395-1G
APOSPC302395-25GEA 1425 SEK
APOSPC302395-25G APOSPC302395-5G APOSPC302395-1G
3-Chloro-4-(trifluoromethyl)benzoic acid 97%
3-Chloro-4-(trifluoromethyl)benzoic acid
Formel: C₈H₄ClF₃O₂
MW: 224.57 g/mol
Smältpunkt: 158…161 °C
Storage Temperature: Omgivande
MDL Number: MFCD04972755
CAS nummer: 115754-20-6