2-Fluoro-5-methoxy-3-(trifluoromethyl)phenylacetic acid 98%
Leverantör: Apollo Scientific
Varning
PC302387-1G PC302387-5G
APOSPC302387-1GEA 1250 SEK
APOSPC302387-1G APOSPC302387-5G
2-Fluoro-5-methoxy-3-(trifluoromethyl)phenylacetic acid 98%
2-Fluoro-5-methoxy-3-(trifluoromethyl)phenylacetic acid
Formel: C₁₀H₈F₄O₃
Storage Temperature: Omgivande
MDL Number: MFCD22201046
CAS nummer: 1373920-85-4