4-Ethoxy-3-(trifluoromethyl)benzaldehyde 97%
Leverantör: Apollo Scientific
Varning
PC302333-25G PC302333-5G PC302333-1G
APOSPC302333-25GEA 6900 SEK
APOSPC302333-25G APOSPC302333-5G APOSPC302333-1G
4-Ethoxy-3-(trifluoromethyl)benzaldehyde 97%
4-Ethoxy-3-(trifluoromethyl)benzaldehyde
Formel: C₁₀H₉F₃O₂
Storage Temperature: Omgivande
MDL Number: MFCD06247417
CAS nummer: 883541-11-5