4-Chloro-2,6-difluoroaniline 98%

Leverantör: Apollo Scientific
Fara

PC302322-5G PC302322-10G
APOSPC302322-5GEA 380 SEK
APOSPC302322-5G APOSPC302322-10G
4-Chloro-2,6-difluoroaniline 98%
4-Chloro-2,6-difluoroaniline
Formel: C₆H₄ClF₂N
MW: 163,55 g/mol
Smältpunkt: 47…50 °C
Storage Temperature: Omgivande
MDL Number: MFCD03426173
CAS nummer: 69411-06-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR