2-Fluoro-4-(trifluoromethoxy)benzoyl chloride 98%
Leverantör: Apollo Scientific
Fara
PC302240-1G PC302240-5G PC302240-10G
APOSPC302240-1GEA 1700 SEK
APOSPC302240-1G APOSPC302240-5G APOSPC302240-10G
2-Fluoro-4-(trifluoromethoxy)benzoyl chloride 98%
2-Fluoro-4-(trifluoromethoxy)benzoyl chloride
Formel: C₈H₃ClF₄O₂
MW: 242.55 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD16652499
UN: 3265
ADR: 8 III