3-(2,4-Difluorophenyl)propionic acid 97%
Leverantör: Apollo Scientific
Varning
PC302182-5G PC302182-1G
APOSPC302182-5GEA 685 SEK
APOSPC302182-5G APOSPC302182-1G
3-(2,4-Difluorophenyl)propionic acid 97%
3-(2,4-Difluorophenyl)propionic acid
Formel: C₉H₈F₂O₂
MW: 186.16 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD04116052
CAS nummer: 134672-70-1