3-Chloro-5-(pentafluorothio)benzaldehyde 97%
Leverantör: Apollo Scientific
|
Varning
|
PC302048-1G
PC302048-5G
PC302048-10G
APOSPC302048-1GEA
4125
SEK
APOSPC302048-1G
APOSPC302048-5G
APOSPC302048-10G
3-Chloro-5-(pentafluorothio)benzaldehyde 97%
3-Chloro-5-(pentafluorothio)benzaldehyde
Formel:
C₇H₄ClF₅OS MW: 266,61 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD16652412 CAS nummer: 1240257-88-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...