1,4-Dibromo-2-(trifluoromethoxy)benzene 97%
Leverantör: Apollo Scientific
PC302014-25G PC302014-5G PC302014-1G
APOSPC302014-25GEA 2250 SEK
APOSPC302014-25G APOSPC302014-5G APOSPC302014-1G
1,4-Dibromo-2-(trifluoromethoxy)benzene 97%
1,4-Dibromo-2-(trifluoromethoxy)benzene
Formel: C₇H₃Br₂F₃O
MW: 319.90 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00179335
CAS nummer: 175278-13-4