Dimethyl tetrafluorosuccinate 98%
Leverantör: Apollo Scientific
Varning
Synonyms: Perfluorosuccinic Acid Dimethyl Ester, Tetrafluorobutanedioic Acid Dimethyl Ester, Dimethyl Tetrafluorobutanedioate, Tetrafluorosuccinic Acid Dimethyl Ester, Dimethyl Perfluorosuccinate
PC3016T-25G PC3016T-5G PC3016T-100G
APOSPC3016T-25GEA 1475 SEK
APOSPC3016T-25G APOSPC3016T-5G APOSPC3016T-100G
Dimethyl tetrafluorosuccinate 98%
Dimethyl tetrafluorosuccinate
Formel: C₆H₆F₄O₄
MW: 218,10 g/mol
Kokpunkt: 177…178 °C
Densitet: 1,41 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00043947
CAS nummer: 356-36-5