3-Carboxy-4-fluorophenylboronic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 3-Carboxy-4-fluorophenylboronic Acid, 3-Carboxy-4-fluorobenzeneboronic acid
PC2998-25G PC2998-5G PC2998-100G
APOSPC2998-25GEA 1075 SEK
APOSPC2998-25G APOSPC2998-5G APOSPC2998-100G
3-Carboxy-4-fluorophenylboronic acid
3-Carboxy-4-fluorophenylboronic acid
Formel: C₇H₆BFO₄
MW: 183.93 g/mol
Smältpunkt: 217…219 °C
Storage Temperature: Omgivande
MDL Number: MFCD03095129
CAS nummer: 872460-12-3