3-Amino-2-fluoro-6-methylpyridine
Leverantör: Apollo Scientific
Varning
PC2987-1G PC2987-5G
APOSPC2987-1GEA 175 SEK
APOSPC2987-1G APOSPC2987-5G
3-Amino-2-fluoro-6-methylpyridine
3-Amino-2-fluoro-6-methylpyridine
Formel: C₆H₇FN₂
Storage Temperature: Omgivande
MDL Number: MFCD03095221
CAS nummer: 374633-34-8