3,4-Difluorophenyl isocyanate 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 3,4-Difluorophenylisocyanate, Isocyanic acid 3,4-Difluorophenyl ester
PC2875K-25G PC2875K-5G PC2875K-1G
APOSPC2875K-25GEA 1550 SEK
APOSPC2875K-25G APOSPC2875K-5G APOSPC2875K-1G
3,4-Difluorophenyl isocyanate 97%
3,4-Difluorophenyl isocyanate
  • May form small amount of sediment.
Formel: C₇H₃F₂NO
MW: 155.1 g/mol
Kokpunkt: 164 °C
Densitet: 1.326 g/cm³ (25 °C)
Flampunkt: 63 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00065711
CAS nummer: 42601-04-7
UN: 2206
ADR: 6.1 III