2,5-Difluorophenyl isocyanate 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 1,4-Difluoro-2-isocyanatobenzene
PC2875I-1G PC2875I-5G
APOSPC2875I-1GEA 245 SEK
APOSPC2875I-1G APOSPC2875I-5G
2,5-Difluorophenyl isocyanate 97%
2,5-Difluorophenyl isocyanate
  • May form small amount of sediment.
Formel: C₇H₃F₂NO
MW: 155,1 g/mol
Kokpunkt: 78 °C/33mmHg
Densitet: 1,323 g/cm³ (25 °C)
Flampunkt: 56 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00013843
CAS nummer: 39718-32-6
UN: 3080
ADR: 6.1+3,II