2,5-Difluorophenylhydrazinium chloride 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 1-(2,5-Difluorophenyl)hydrazine hydrochloride, 2,5-Difluorophenylhydrazine hydrochloride
PC2874X-10G PC2874X-5G
APOSPC2874X-10GEA 1125 SEK
APOSPC2874X-10G APOSPC2874X-5G
2,5-Difluorophenylhydrazinium chloride 97%
2,5-Difluorophenylhydrazinium chloride
Formel: C₆H₇ClF₂N₂
MW: 180,58 g/mol
Smältpunkt: 210…211 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00013385
CAS nummer: 175135-73-6