2,4-Difluorophenylhydrazinium chloride, Technical Grade
Leverantör: Apollo Scientific
Varning
Synonyms: 2,4-Difluorophenylhydrazine hydrochloride, 1-(2,4-Difluorophenyl)hydrazine hydrochloride
PC2874V-100G PC2874V-25G PC2874V-5G
APOSPC2874V-100GEA 910 SEK
APOSPC2874V-100G APOSPC2874V-25G APOSPC2874V-5G
2,4-Difluorophenylhydrazinium chloride, Technical Grade
2,4-Difluorophenylhydrazinium chloride
Formel: C₆H₇ClF₂N₂
MW: 180.58 g/mol
Smältpunkt: 250…260 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00013383
CAS nummer: 51523-79-6