2-Bromo-6-(trifluoromethyl)aniline 98%
Leverantör: Apollo Scientific
Varning
PC2857-1G PC2857-5G PC2857-10G
APOSPC2857-1GEA 1500 SEK
APOSPC2857-1G APOSPC2857-5G APOSPC2857-10G
2-Bromo-6-(trifluoromethyl)aniline 98%
2-Bromo-6-(trifluoromethyl)aniline
Formel: C₇H₅BrF₃N
Storage Temperature: Kylskåp
MDL Number: MFCD08741383
CAS nummer: 58458-13-2