2-Fluoro-4-(hydroxymethyl)benzonitrile

Leverantör: Apollo Scientific
Varning

PC2806-1G PC2806-5G
APOSPC2806-1GEA 1200 SEK
APOSPC2806-1G APOSPC2806-5G
2-Fluoro-4-(hydroxymethyl)benzonitrile
2-Fluoro-4-(hydroxymethyl)benzonitrile
Formel: C₈H₆FNO
MW: 151,1400 g/mol
Smältpunkt: 66…68 °C
Storage Temperature: Omgivande
MDL Number: MFCD08236861
CAS nummer: 222978-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR