1-(4-Fluorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid 95%
Leverantör: Apollo Scientific
PC28011-1G PC28011-5G
APOSPC28011-1GEA 2750 SEK
APOSPC28011-1G APOSPC28011-5G
1-(4-Fluorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid 95%
1-(4-Fluorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid
Formel: C₁₂H₁₁FN₂O₂
Storage Temperature: Omgivande
MDL Number: MFCD02083021
CAS nummer: 288251-63-8