trans-3,4-Difluorocinnamic acid 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 3,4-Difluorocinnamic acid
PC2764-25G PC2764-5G PC2764-100G
APOSPC2764-25GEA 345 SEK
APOSPC2764-25G APOSPC2764-5G APOSPC2764-100G
trans-3,4-Difluorocinnamic acid 97%
trans-3,4-Difluorocinnamic acid
Formel: C₉H₆F₂O₂
MW: 184,14 g/mol
Smältpunkt: 194…198 °C
Storage Temperature: Omgivande
MDL Number: MFCD00010320
CAS nummer: 112897-97-9