(3,4-Difluorophenyl)hydrazine hydrochloride 97%
Leverantör: Apollo Scientific
Varning
Synonyms: (3,4-Difluorophenyl)hydrazine xhydrochloride, 3,4-Difluorophenylhydrazinium chloride, 1-(3,4-Difluorophenyl)hydrazine hydrochloride
PC2720-5G PC2720-25G PC2720-100G
APOSPC2720-5GEA 260 SEK
APOSPC2720-5G APOSPC2720-25G APOSPC2720-100G
(3,4-Difluorophenyl)hydrazine hydrochloride 97%
(3,4-Difluorophenyl)hydrazine hydrochloride
Formel: C₆H₇ClF₂N₂
MW: 180.59 g/mol
Smältpunkt: ∼ 230 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD03094170
CAS nummer: 875664-54-3