3-Fluoro-2-iodopyridine
Leverantör: Apollo Scientific
Varning
PC2675-1G PC2675-250MG PC2675-5G
APOSPC2675-1GEA 1075 SEK
APOSPC2675-1G APOSPC2675-250MG APOSPC2675-5G
3-Fluoro-2-iodopyridine
3-Fluoro-2-iodopyridine
Formel: C₅H₃FIN
Storage Temperature: Omgivande
MDL Number: MFCD03095288
CAS nummer: 146141-04-0