2-Amino-3-(trifluoromethyl)pyridine 98%

Leverantör: Apollo Scientific
Fara

PC2512-25G PC2512-5G
APOSPC2512-25GEA 1150 SEK
APOSPC2512-25G APOSPC2512-5G
2-Amino-3-(trifluoromethyl)pyridine 98%
2-Amino-3-(trifluoromethyl)pyridine
Formel: C₆H₅F₃N₂
Smältpunkt: 66…72 °C
Storage Temperature: Omgivande
MDL Number: MFCD04972538
CAS nummer: 183610-70-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR