Trifluorothymidine 98%
Leverantör: Apollo Scientific
|
Varning
|
PC2508-1G
PC2508-250MG
APOSPC2508-1GEA
0
SEK
APOSPC2508-1G
APOSPC2508-250MG
Trifluorothymidine 98%
Trifluorothymidine
Formel:
C₁₀H₁₁F₃N₂O₅ Smältpunkt: 190…193 °C Storage Temperature: Frys |
MDL Number:
MFCD00006534 CAS nummer: 70-00-8 EINECS: 200-722-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...