2,3-Dibromo-1,1,3,3-tetrafluoropropene 97%
Leverantör: Apollo Scientific
PC2322G-1G PC2322G-5G
APOSPC2322G-1GEA 1300 SEK
APOSPC2322G-1G APOSPC2322G-5G
2,3-Dibromo-1,1,3,3-tetrafluoropropene 97%
2,3-Dibromo-1,1,3,3-tetrafluoropropene
Precursor to tetrafluoroallene.
Formel: C₃Br₂F₄
Storage Temperature: Omgivande
MDL Number: MFCD00236663
CAS nummer: 666-40-0