2-Chloro-6-(trifluoromethyl)nicotinic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Chloro-6-(trifluoromethyl)pyridine-3-carboxylic acid
PC2223-1G PC2223-25G PC2223-5G
APOSPC2223-1GEA 260 SEK
APOSPC2223-1G APOSPC2223-25G APOSPC2223-5G
2-Chloro-6-(trifluoromethyl)nicotinic acid
2-Chloro-6-(trifluoromethyl)nicotinic acid
Formel: C₇H₃ClF₃NO₂
MW: 225.56 g/mol
Smältpunkt: 120…122 °C
Storage Temperature: Omgivande
MDL Number: MFCD01862658
CAS nummer: 280566-45-2