Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate
Leverantör: Apollo Scientific
Synonyms: Bis(1,5-cyclooctadiene)iridium (I) tetrafluoroborate
PC2197-500MG PC2197-1G
APOSPC2197-500MGEA 6625 SEK
APOSPC2197-500MG APOSPC2197-1G
Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate
Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate
Formel: C₁₆H₂₄BF₄Ir
MW: 495.383 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00079664
CAS nummer: 35138-23-9