2-Fluoroisonicotinic acid 98%
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
2-Fluoropyridine-4-carboxylic acid
PC2057-1G
APOSPC2057-1GEA
260
SEK
APOSPC2057-1G
2-Fluoroisonicotinic acid 98%
2-Fluoroisonicotinic acid
Formel:
C₆H₄FNO₂ MW: 141.10 g/mol Smältpunkt: 191…200 °C (dec.) Storage Temperature: Omgivande |
MDL Number:
MFCD02181194 CAS nummer: 402-65-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...