2-Chloro-5-(trifluoromethyl)pyridin-3-amine 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 3-Amino-2-chloro-5-(trifluoromethyl)pyridine
PC201291-10G PC201291-5G PC201291-1G
APOSPC201291-10GEA 2625 SEK
APOSPC201291-10G APOSPC201291-5G APOSPC201291-1G
2-Chloro-5-(trifluoromethyl)pyridin-3-amine 98%
2-Chloro-5-(trifluoromethyl)pyridin-3-amine
Formel: C₆H₄ClF₃N₂
MW: 196.557 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD07375383
CAS nummer: 72587-18-9