(R)-Boroleucine-(1S,2S,3R,5S)-(+)-pinanediol ester trifluoroacetate 98%
Leverantör: Apollo Scientific
Varning
Synonyms: (R)-3-Methyl-1-((3aS,4S,6S,7aR)-3a,5,5-trimethylhexahydro-4,6-methanobenzo[d][1,3,2]dioxaborol-2-yl)butan-1-amine 2,2,2-trifluoroacetate, Bortezomib Amino R Isomer (TFA salt), (R)-BoroLeu-(+)-Pinanediol trifluoroacetate
PC200351-1G PC200351-5G
APOSPC200351-1GEA 945 SEK
APOSPC200351-1G APOSPC200351-5G
(R)-Boroleucine-(1S,2S,3R,5S)-(+)-pinanediol ester trifluoroacetate 98%
(R)-Boroleucine-(1S,2S,3R,5S)-(+)-pinanediol ester trifluoroacetate
Formel: C₁₇H₂₉BF₃NO₄
MW: 379,23 g/mol
Storage Temperature: Frys
MDL Number: MFCD10566030
CAS nummer: 179324-87-9