Methyl-2,6-dichloro-5-(trifluoromethyl)-4-pyrimidinecarboxylate
Leverantör: Apollo Scientific
PC200324-500MG
APOSPC200324-500MGEA
4850
SEK
APOSPC200324-500MG
Methyl-2,6-dichloro-5-(trifluoromethyl)-4-pyrimidinecarboxylate
Methyl-2,6-dichloro-5-(trifluoromethyl)-4-pyrimidinecarboxylate
Formel:
C₇H₃Cl₂F₃N₂O₂ MW: 275.02 g/mol Smältpunkt: 57…58 °C Storage Temperature: Omgivande |
MDL Number:
MFCD12922697 CAS nummer: 1190198-37-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...