[2,6-Bis(trifluoromethyl)pyridin-4-yl]methanol 95+%
Leverantör: Apollo Scientific
Synonyms: 2,6-Bis(trifluoromethyl)-4-(hydroxymethyl)pyridine
PC200301-1G PC200301-500MG
APOSPC200301-1GEA 11575 SEK
APOSPC200301-1G APOSPC200301-500MG
[2,6-Bis(trifluoromethyl)pyridin-4-yl]methanol 95+%
[2,6-Bis(trifluoromethyl)pyridin-4-yl]methanol
Formel: C₈H₅F₆NO
MW: 245.13 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD20921614
CAS nummer: 1353878-03-1